2,6-Diaminopimelic acid 96%
Supplier: Apollo Scientific
Synonyms: 1,5-Diaminoheptanedioic Acid, 2,6-Diaminoheptanedioic acid
OR480250-25G OR480250-5G
APOSOR480250-25GEA 941 EUR
APOSOR480250-25G APOSOR480250-5G
2,6-Diaminopimelic acid 96%
2,6-Diaminopimelic acid
Formula: C₇H₁₄N₂O₄
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002637
CAS Number: 583-93-7