4-(3-Isopropyl[1,2,4]oxadiazol-5-yl)piperidine-1-carboxylic acid tert-butyl ester 95%
Supplier: Apollo Scientific
OR480269-10G OR480269-1G OR480269-5G
APOSOR480269-10GEA 919 EUR
APOSOR480269-10G APOSOR480269-1G APOSOR480269-5G
4-(3-Isopropyl[1,2,4]oxadiazol-5-yl)piperidine-1-carboxylic acid tert-butyl ester 95%
4-(3-Isopropyl[1,2,4]oxadiazol-5-yl)piperidine-1-carboxylic acid tert-butyl ester
Formula: C₁₅H₂₅N₃O₃
MW: 295.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226634
CAS Number: 913264-42-3