(S)-2-Boc-aminomethylpyrrolidine 95%
Supplier: Apollo Scientific
Synonyms: (S)-tert-butyl pyrrolidin-2-ylmethylcarbamate, (S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate
OR480535-25G OR480535-5G
APOSOR480535-25GEA 725 EUR
APOSOR480535-25G APOSOR480535-5G
(S)-2-Boc-aminomethylpyrrolidine 95%
(S)-2-Boc-aminomethylpyrrolidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796572
CAS Number: 141774-70-1