2-Methyl-4-isothiazolin-3-one hydrochloride 98%
Supplier: Apollo Scientific
Synonyms: Methylisothiazolinone hydrochloride, MIT
OR480577-1G OR480577-5G OR480577-25G
APOSOR480577-1GEA 21.9 EUR
APOSOR480577-1G APOSOR480577-5G APOSOR480577-25G
2-Methyl-4-isothiazolin-3-one hydrochloride 98%
2-Methyl-4-isothiazolin-3-one hydrochloride
Formula: C₄H₆ClNOS
MW: 151.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06804636
CAS Number: 26172-54-3