(R)-(+)-Epoxystyrene 95%
Supplier: Apollo Scientific
Danger
Synonyms: (R)-(+)-Styrene oxide, (R)-Styrene oxide
OR480595-100G OR480595-25G
APOSOR480595-100GEA 720 EUR
APOSOR480595-100G APOSOR480595-25G
(R)-(+)-Epoxystyrene 95%
(R)-(+)-Epoxystyrene
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 192…195 °C/760mmHg
Melting Pt: –37 °C
Density: 1.053 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066210
CAS Number: 20780-53-4
UN: 2810
ADR: 6.1 III