4-Methyl-1H-pyrazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: Fomepizole
OR480607-100G OR480607-25G OR480607-500G
APOSOR480607-100GEA 219 EUR
APOSOR480607-100G APOSOR480607-25G APOSOR480607-500G
4-Methyl-1H-pyrazole 98%
4-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 99…100 °C/6mmHg
Density: 0.99 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005245
CAS Number: 7554-65-6
UN: 1760
ADR: 8,III