3-Methoxy-p-toluidine 95%
Supplier: Apollo Scientific
Synonyms:
1-Amino-3-methoxy-4-methylbenzene, 4-Amino-2-methoxytoluene, 3-Methoxy-4-methylaniline, 4-Methyl-m-anisidine, 5-Amino-2-methylanisole
OR480640-25G
OR480640-5G
APOSOR480640-25GEA
207
EUR
APOSOR480640-25G
APOSOR480640-5G
3-Methoxy-p-toluidine 95%
3-Methoxy-p-toluidine
Formula:
C₈H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD00025371 CAS Number: 16452-01-0 |