(1S,2R)-cis-2-Aminocyclohexanol hydrochloride 98%
Supplier: Apollo Scientific
Synonyms: (1S,2R)-2-Aminocyclohexanol hydrochloride
OR480654-1G OR480654-5G
APOSOR480654-1GEA 21.9 EUR
APOSOR480654-1G APOSOR480654-5G
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride 98%
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride
Formula: C₆H₁₄ClNO
MW: 151.6345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11618003
CAS Number: 200352-28-9