Pempidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2,2,6,6-Pentamethylpiperidine
OR480665-25G
APOSOR480665-25GEA 155 EUR
APOSOR480665-25G
Pempidine 97%
Pempidine
Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Boiling Pt: 187…188 °C
Density: 0.858 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00006493
CAS Number: 79-55-0
UN: 1992
ADR: 3+6.1,III