4-Benzyloxy-2-chloropyrimidine 95%

Supplier: Apollo Scientific

APOSOR480782-1GEA 209 EUR
APOSOR480782-1G
4-Benzyloxy-2-chloropyrimidine 95%
4-Benzyloxy-2-chloropyrimidine
Formula: C₁₁H₉ClN₂O
MW: 220,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798233
CAS Number: 108381-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR