Oxolane-3-carbonitrile
Supplier: Apollo Scientific
Synonyms: Tetrahydrofuran-3-carbonitrile
OR480787-1G OR480787-5G
APOSOR480787-1GEA 84.1 EUR
APOSOR480787-1G APOSOR480787-5G
Oxolane-3-carbonitrile
Oxolane-3-carbonitrile
Formula: C₅H₇NO
MW: 97.1152 g/mol
Storage Temperature: Ambient
CAS Number: 14631-44-8