1-Methylimidazole-2-sulphonyl chloride 95%
Supplier: Apollo Scientific
OR480795-1G
OR480795-5G
APOSOR480795-1GEA
226
EUR
APOSOR480795-1G
APOSOR480795-5G
1-Methylimidazole-2-sulphonyl chloride 95%
1-Methylimidazole-2-sulphonyl chloride
Formula:
C₄H₅ClN₂O₂S Melting Pt: 66…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD16621450 CAS Number: 55694-81-0 UN: 3261 ADR: 8,III |