Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Supplier: Apollo Scientific
Warning
Synonyms: Lithium PTMB phosphinate, LAP, Phenyl(2,4,6-trimethylbenzoyl)phosphinic acid lithium salt, Lithium Phenyl(2,4,6-trimethylbenzoyl)phosphinate
OR48106-1G OR48106-500MG OR48106-5G
APOSOR48106-1GEA 145 EUR
APOSOR48106-1G APOSOR48106-500MG APOSOR48106-5G
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP)
Formula: C₁₆H₁₆LiO₃P
MW: 294.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29472537
CAS Number: 85073-19-4