3-Amino-4-methylbenzonitrile 98%
Supplier: Apollo Scientific
Warning
OR4824-25G OR4824-5G OR4824-100G
APOSOR4824-25GEA 30.6 EUR
APOSOR4824-25G APOSOR4824-5G APOSOR4824-100G
3-Amino-4-methylbenzonitrile 98%
3-Amino-4-methylbenzonitrile
Formula: C₈H₈N₂
MW: 132.1600 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD04039781
CAS Number: 60710-80-7