Diethyl (3-bromopropyl)phosphonate 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Bromopropyl)phosphonic Acid Diethyl Ester
OR48243-1G OR48243-5G OR48243-10G
APOSOR48243-1GEA 24.8 EUR
APOSOR48243-1G APOSOR48243-5G APOSOR48243-10G
Diethyl (3-bromopropyl)phosphonate 98%
Diethyl (3-bromopropyl)phosphonate
Formula: C₇H₁₆BrO₃P
MW: 259.08 g/mol
Boiling Pt: 94…95 °C/2.25mmHg
Density: 1.348 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00800966
CAS Number: 1186-10-3