Pyrimidin-2-ylmethanamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Aminomethyl)pyrimidine hydrochloride
OR48262-25G OR48262-5G
APOSOR48262-25GEA 132 EUR
APOSOR48262-25G APOSOR48262-5G
Pyrimidin-2-ylmethanamine hydrochloride
Pyrimidin-2-ylmethanamine hydrochloride
Formula: C5H8ClN3
MW: 145.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062208
CAS Number: 372118-67-7