3-Methoxybenzonitrile 99%
Supplier: Apollo Scientific
Warning
OR4831-100G OR4831-25G
APOSOR4831-100GEA 72.5 EUR
APOSOR4831-100G APOSOR4831-25G
3-Methoxybenzonitrile 99%
3-Methoxybenzonitrile
Formula: C₈H₇NO
Boiling Pt: 110…112 °C/12mmHg
Melting Pt: 22…24 °C
Density: 1.089 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00001801
CAS Number: 1527-89-5
EINECS: 216-201-3
UN: 3439
ADR: 6.1,III