3-Aminothiophene-2-carboxamide 98%
Supplier: Apollo Scientific
Warning
OR4837-1G OR4837-250MG
APOSOR4837-1GEA 75.7 EUR
APOSOR4837-1G APOSOR4837-250MG
3-Aminothiophene-2-carboxamide 98%
3-Aminothiophene-2-carboxamide
Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00052593
CAS Number: 147123-47-5