tert-Butyl pyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-butyl pyrrolidine-1-carboxylate, pyrrolidine-1-carboxylic acid tert-butyl ester, N-BOC-Pyrrolidine
OR4852-1G OR4852-5G
APOSOR4852-1GEA 21.9 EUR
APOSOR4852-1G APOSOR4852-5G
tert-Butyl pyrrolidine-1-carboxylate 97%
tert-Butyl pyrrolidine-1-carboxylate
Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 80 °C/0.2mmHg
Density: 0.977 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00216581
CAS Number: 86953-79-9