7-Ethoxy-4-methylcoumarin 98%
Supplier: Apollo Scientific
Warning
OR4853-100G OR4853-25G OR4853-5G
APOSOR4853-100GEA 116 EUR
APOSOR4853-100G APOSOR4853-25G APOSOR4853-5G
7-Ethoxy-4-methylcoumarin 98%
7-Ethoxy-4-methylcoumarin
Formula: C₁₂H₁₂O₃
MW: 204.23 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00016971
CAS Number: 87-05-8
EINECS: 201-721-5