6-Chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Warning
OR4874-100MG OR4874-1G OR4874-5G
APOSOR4874-100MGEA 23.2 EUR
APOSOR4874-100MG APOSOR4874-1G APOSOR4874-5G
6-Chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
6-Chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269625
CAS Number: 138642-96-3