Uracil
Supplier: Apollo Scientific
Warning
Synonyms: Pyrimidine-2,4(1H,3H)-dione
OR4885-250G OR4885-100G OR4885-25G
APOSOR4885-250GEA 50.9 EUR
APOSOR4885-250G APOSOR4885-100G APOSOR4885-25G
Uracil
Uracil
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 379
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985