3-Chloroaniline
Supplier: Apollo Scientific
Danger
OR4892-100G OR4892-500G
APOSOR4892-100GEA 21.9 EUR
APOSOR4892-100G APOSOR4892-500G
3-Chloroaniline
3-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 95…96 °C/11mmHg
Melting Pt: –11 °C
Density: 1.206 g/cm³ (25 °C)
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00007765
CAS Number: 108-42-9
EINECS: 203-581-0
UN: 2019
ADR: 6.1,II