4-Bromobenzylamine 98%
Supplier: Apollo Scientific
Danger
OR49010-100G OR49010-25G OR49010-5G
APOSOR49010-100GEA 212 EUR
APOSOR49010-100G APOSOR49010-25G APOSOR49010-5G
4-Bromobenzylamine 98%
4-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 110…112 °C/30mmHg
Melting Pt: 25 °C
Density: 1.473 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047931
CAS Number: 3959-07-7
EINECS: 223-559-4
UN: 3259
ADR: 8 II