2-Chloro-N,N-dimethylacetamide 97%
Supplier: Apollo Scientific
OR49017-100G OR49017-25G
APOSOR49017-100GEA 408 EUR
APOSOR49017-100G APOSOR49017-25G
2-Chloro-N,N-dimethylacetamide 97%
2-Chloro-N,N-dimethylacetamide
Formula: C₄H₈ClNO
MW: 121.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039360
CAS Number: 2675-89-0