4-(Dimethylamino)cyclohexanone
Supplier: Apollo Scientific
Danger
OR49023-2G OR49023-5G
APOSOR49023-2GEA 183 EUR
APOSOR49023-2G APOSOR49023-5G
4-(Dimethylamino)cyclohexanone
4-(Dimethylamino)cyclohexanone
Formula: C₈H₁₅NO
MW: 141.2100 g/mol
Boiling Pt: 213.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037892
CAS Number: 40594-34-1
UN: 2735
ADR: 8 III