Crotyl alcohol 96%
Supplier: Apollo Scientific
Synonyms: 2-Buten-1-ol, 1-Hydroxy-2-butene
OR49073-25ML OR49073-100ML
APOSOR49073-25MLEA 201 EUR
APOSOR49073-25ML APOSOR49073-100ML
Crotyl alcohol 96%
Crotyl alcohol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 120
Storage Temperature: Ambient
MDL Number: MFCD00002923
CAS Number: 6117-91-5
UN: 1987
ADR: 3 III
Merck Index: 13,02629