4,5-Dichloro-o-phenylenediamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine
OR4932-100G OR4932-25G OR4932-5G
APOSOR4932-100GEA 455 EUR
APOSOR4932-100G APOSOR4932-25G APOSOR4932-5G
4,5-Dichloro-o-phenylenediamine 98%
4,5-Dichloro-o-phenylenediamine
Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 305
Melting Pt: 158…163 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007723
CAS Number: 5348-42-5