5-Isoxazol-5-ylthiophene-2-sulphonyl chloride
Supplier: Apollo Scientific
Danger
OR4941-1G OR4941-250MG OR4941-5G
APOSOR4941-1GEA 143 EUR
APOSOR4941-1G APOSOR4941-250MG APOSOR4941-5G
5-Isoxazol-5-ylthiophene-2-sulphonyl chloride
5-Isoxazol-5-ylthiophene-2-sulphonyl chloride
Formula: C₇H₄ClNO₃S₂
MW: 249.7 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02187482
CAS Number: 551930-53-1
UN: 3261
ADR: 8,III