4-Isopropylaniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: p-Cumidine, 4-Aminocumene
OR4957-100G OR4957-5G OR4957-10G OR4957-25G
APOSOR4957-100GEA 0 EUR
APOSOR4957-100G APOSOR4957-5G APOSOR4957-10G APOSOR4957-25G
4-Isopropylaniline 98%
4-Isopropylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 224…228 °C
Melting Pt: –63 °C
Density: 0.989 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7
EINECS: 202-797-2
UN: 2922
ADR: 8+6.1,III