4-Chloro-3-nitrobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR4977-10G OR4977-50G OR4977-25G OR4977-100G
APOSOR4977-10GEA 26.1 EUR
APOSOR4977-10G APOSOR4977-50G APOSOR4977-25G APOSOR4977-100G
4-Chloro-3-nitrobenzaldehyde 97%
4-Chloro-3-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00007078
CAS Number: 16588-34-4