3-Chlorobenzaldehyde
Supplier: Apollo Scientific
Warning
OR4978-500G
APOSOR4978-500GEA 92.8 EUR
APOSOR4978-500G
3-Chlorobenzaldehyde
3-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 16…18 °C
Density: 1.239 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9