4-Bromobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR4979-100G OR4979-25G
APOSOR4979-100GEA 46.4 EUR
APOSOR4979-100G APOSOR4979-25G
4-Bromobenzaldehyde 98%
4-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 66…68 °C/2mmHg
Melting Pt: 55…58 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0