4-Ethoxybenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR4980-100G OR4980-25G OR4980-500G
APOSOR4980-100GEA 144 EUR
APOSOR4980-100G APOSOR4980-25G APOSOR4980-500G
4-Ethoxybenzaldehyde 98%
4-Ethoxybenzaldehyde
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 255 °C
Melting Pt: 13…14 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00003388
CAS Number: 10031-82-0
EINECS: 233-093-3