5-Nitroanthranilic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-nitrobenzoic acid
OR4994-100G OR4994-25G OR4994-500G
APOSOR4994-100GEA 72.5 EUR
APOSOR4994-100G APOSOR4994-25G APOSOR4994-500G
5-Nitroanthranilic acid 98%
5-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00017039
CAS Number: 616-79-5