4-Iodobenzoic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: p-Iodobenzoic acid
OR4999-100G OR4999-25G
APOSOR4999-100GEA 43.5 EUR
APOSOR4999-100G APOSOR4999-25G
4-Iodobenzoic acid 98%
4-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319
Melting Pt: 270…273 °C (dec.)
Density: 2.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2