2-Nitrobenzonitrile 99%
Supplier: Apollo Scientific
Danger
Synonyms: o-Nitrobenzonitrile
OR5001-25G OR5001-1G OR5001-5G
APOSOR5001-25GEA 72.7 EUR
APOSOR5001-25G APOSOR5001-1G APOSOR5001-5G
2-Nitrobenzonitrile 99%
2-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 165 °C/16mmHg
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007044
CAS Number: 612-24-8
EINECS: 210-301-0
UN: 3439
ADR: 6.1 III