4,4'-Dichlorobenzophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4-Dichlorobenzophenone, 4,4-DBP, Bis(4-chlorophenyl) ketone
OR5010-100G OR5010-250G OR5010-500G
APOSOR5010-100GEA 26.1 EUR
APOSOR5010-100G APOSOR5010-250G APOSOR5010-500G
4,4'-Dichlorobenzophenone 98%
4,4'-Dichlorobenzophenone
Formula: C₁₃H₈Cl₂O
Boiling Pt: 353 °C
Melting Pt: 144…146 °C
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000623
CAS Number: 90-98-2
EINECS: 202-030-1