4-Nitrobenzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Nitrophenyl)methanol
OR5015-100G
OR5015-1KG
OR5015-500G
APOSOR5015-100GEA
21.9
EUR
APOSOR5015-100G
APOSOR5015-1KG
APOSOR5015-500G
4-Nitrobenzyl alcohol 98%
4-Nitrobenzyl alcohol
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Boiling Pt: 185 °C/12mmHg Melting Pt: 92…95 °C Flash Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007376 CAS Number: 619-73-8 EINECS: 210-611-6 |