3-Nitrobenzyl alcohol 99%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Nitrophenyl)methanol
OR5016-100G OR5016-25G
APOSOR5016-100GEA 53.8 EUR
APOSOR5016-100G APOSOR5016-25G
3-Nitrobenzyl alcohol 99%
3-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 175…180 °C/3mmHg
Melting Pt: 30…32 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007273
CAS Number: 619-25-0