3-Chlorobenzyl alcohol 99%
Supplier: Apollo Scientific
Warning
OR5019-10G OR5019-100G OR5019-25G
APOSOR5019-10GEA 21.9 EUR
APOSOR5019-10G APOSOR5019-100G APOSOR5019-25G
3-Chlorobenzyl alcohol 99%
3-Chlorobenzyl alcohol
Formula: C₇H₇ClO
Boiling Pt: 237 °C
Density: 1.211 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004632
CAS Number: 873-63-2
EINECS: 212-847-5