3-Chlorobenzylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: (3-Chlorophenyl)methanamine
OR5026-25G
APOSOR5026-25GEA 50.9 EUR
APOSOR5026-25G
3-Chlorobenzylamine 98%
3-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 110…112 °C/17mmHg
Density: 1.159 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00040752
CAS Number: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8,II