2-(Aminomethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine
OR5031-100G OR5031-25G
APOSOR5031-100GEA 43.5 EUR
APOSOR5031-100G APOSOR5031-25G
2-(Aminomethyl)pyridine 98%
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 202…204 °C
Melting Pt: –20 °C
Density: 1.049 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5
UN: 2735
ADR: 8,II