4-Phenylcyclohexanone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane
OR5040-100G OR5040-25G OR5040-5G
APOSOR5040-100GEA 108 EUR
APOSOR5040-100G APOSOR5040-25G APOSOR5040-5G
4-Phenylcyclohexanone 98%
4-Phenylcyclohexanone
Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 158…160 °C/16mmHg
Melting Pt: 73…80 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1
EINECS: 225-517-0