2-Acetylpyridine 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone
OR5045-100G OR5045-500G OR5045-250G
APOSOR5045-100GEA 132 EUR
APOSOR5045-100G APOSOR5045-500G APOSOR5045-250G
2-Acetylpyridine 97+%
2-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 188…190 °C
Melting Pt: 44477 °C
Density: 1.082 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006303
CAS Number: 1122-62-9
EINECS: 214-355-6