Thiophene-2-carbaldehyde 97%
Supplier: Apollo Scientific
Warning
OR5053-100G OR5053-25G OR5053-250G
APOSOR5053-100GEA 33.5 EUR
APOSOR5053-100G APOSOR5053-25G APOSOR5053-250G
Thiophene-2-carbaldehyde 97%
Thiophene-2-carbaldehyde
Formula: C₅H₄OS
Boiling Pt: 196…198 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005429
CAS Number: 98-03-3
EINECS: 202-629-8