5-Methylthiophene-2-carboxaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde
OR5055-100G OR5055-10G OR5055-25G
APOSOR5055-100GEA 62.8 EUR
APOSOR5055-100G APOSOR5055-10G APOSOR5055-25G
5-Methylthiophene-2-carboxaldehyde 97%
5-Methylthiophene-2-carboxaldehyde
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 114…116 °C/19mmHg
Density: 1.17 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005434
CAS Number: 13679-70-4
EINECS: 237-178-6