Ethyl thiophene-2-carboxylate 98%
Supplier: Apollo Scientific
Warning
OR5056-25G OR5056-5G
APOSOR5056-25GEA 24.4 EUR
APOSOR5056-25G APOSOR5056-5G
Ethyl thiophene-2-carboxylate 98%
Ethyl thiophene-2-carboxylate
Formula: C₇H₈O₂S
Boiling Pt: 217…218 °C
Density: 1.162 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005436
CAS Number: 2810-04-0
EINECS: 220-554-9