3-Thenoic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid
OR5058-100G OR5058-25G OR5058-5G
APOSOR5058-100GEA 114 EUR
APOSOR5058-100G APOSOR5058-25G APOSOR5058-5G
3-Thenoic acid 98%
3-Thenoic acid
Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00005467
CAS Number: 88-13-1
Merck Index: 12,09415