Naphthacene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3-Benzanthracene, Benz[b]anthracene, Tetracene
OR50973-1G
OR50973-250MG
APOSOR50973-1GEA
161
EUR
APOSOR50973-1G
APOSOR50973-250MG
Naphthacene
Naphthacene
Formula:
C₁₈H₁₂ MW: 228.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003702 CAS Number: 92-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...